missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rho guanine nucleotide exchange factor 18 Protein
Specifications
Specifications
| Zugriffsnummer | NP_001124427.1 |
| Gen-ID (Entrez) | 23370 |
| Name | Rho guanine nucleotide exchange factor 18 Protein |
| Menge | 100 μg |
| Kennzeichnung | Research Use Only |
| Endotoxin-Konzentration | Not determined. |
| Gensymbol | ARHGEF18 |
| Proteinlänge | Partial (see sequence information for more details). |
| Produkttyp | Recombinant Protein |
| Kreuzreaktivität | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?