missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCAN domain-containing protein 1
Click to view available options
Menge:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
200 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Spezifikation
Spezifikation
| Zugriffsnummer | NP_057642.1 |
| Gen-ID (Entrez) | 51282 |
| Name | SCAN domain-containing protein 1 |
| Menge | 1 mg |
| Kennzeichnung | Research Use Only |
| Endotoxin-Konzentration | Not determined. |
| Gensymbol | SCAND1 |
| Proteinlänge | Full Length |
| Produkttyp | Recombinant Protein |
| Kreuzreaktivität | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction