missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Serum Amyloid A1 Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
1 mg
10 μg
100 μg
500 μg
missing translation for 'unitSize'
1 Milligramm
10 Mikrogramm
100 Mikrogramm
500 Mikrogramm
Description
Fully biologically active when compared to standard. The biological activity determined by a chemoattract bioassay using human monocytes is in a concentration range of 10-100 ng/mL.
Specifications
Specifications
| Zugriffsnummer | P0DJI8, P0DJI8 |
| Konzentration | LYOPH |
| Zur Verwendung mit (Anwendung) | PAGE |
| Zusammensetzung | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris-HCl, pH 9.0, 150 mM NaCl. |
| Gen-ID (Entrez) | 6288 |
| Molekulargewicht | 11.7 kDa |
| Menge | 1 mg |
| Lagerungsbedingungen | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Gen-Alias | MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 |
| Gensymbol | SAA1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction