Learn More
enQuireBio™ Recombinant Human SIRT2 Protein
Click to view available options
Menge:
1 μg
5 μg
50 μg
Packungsgröße:
1 Mikrogramm
5 Mikrogramm
50 Mikrogramm
Spezifikation
Spezifikation
| Name | SIRT2 Protein |
| Menge | 1 μg |
| Kennzeichnung | Research Use Only |
| Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Produkttyp | Recombinant Protein |
| Kreuzreaktivität | Human |
| Spezies | E. coli |
| Proteinmarkierung | His |
| Sequenz | MGSSHHHHHH SSGLVPRGSH MDFLRNLFSQ TLSLGSQKER LLDELTLEGV ARYMQSERCR RVICLVGAGI STSAGIPDFR SPSTGLYDNL EKYHLPYPEA IFEISYFKKH PEPFFALAKE LYPGQFKPTI CHYFMRLLKD KGLLLRCYTQ NIDTLERIAG LEQEDLVEAH GTFYTSHCVS ASCRHEYPLS WMKEKIFSEV TPKCEDCQSL VKPDIVFFGE SLPARFFSCM QSDFLKVDLL LVMGTSLQVQ PFASLISKAP LSTPRLLINK EKAGQSDPFL GMIMGLGGGM DFDSKKAYRD VAWLGECDQG CLALAELLGW KKELEDLVRR EHASIDAQSG AGVPNPSTSA SPKKSPPPAK DEARTTEREK PQ |
| Puffer | The SIRT2 protein solution (0.25 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 2mM DTT, 200mM NaCl, 0.5mM EDTA and 30% glycerol. |
| Mehr anzeigen |