Learn More
Novus Biologicals™ Recombinant Human Sonic Hedgehog/Shh Protein
Artikelnummer. 18645245 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
100 μg
5 μg
500 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
5 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
18645245 1 mg 1 Milligramm
18611996 5 μg 5 Mikrogramm
18677215 500 μg 500 Mikrogramm
18650196 100 μg 100 Mikrogramm
4 options
Artikelnummer. 18645245
Marke: Novus Biologicals™ NBP2352651mg
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Protein
Sonic Hedgehog/Shh protein is fully biologically active when compared to standard. The ED50 as determined by inducing alkaline phosphatase production of murine C3H/10T1/2 cells is less than 1 μg/mL, corresponding to a specific activity of > 1.0 x 10^3 IU/mg
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer Q15465, Q15465, Q15465
Zur Verwendung mit (Anwendung) SDS-PAGE, Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl.
Gen-ID (Entrez) 6469
Molekulargewicht MolecularWeight-theroretical: 19.8 kDa
Menge 1 mg
Lagerungsbedingungen Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin-Konzentration Less than 1 EU/μg of Sonic Hedgehog/Shh as determined by LAL method.
Gen-Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Gensymbol SHH
Rekonstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Forschungskategorie Biologically Active Proteins
Reinheits- oder Qualitätsgrad >98 % pure by SDS-PAGE and HPLC
Protein Sonic Hedgehog/Shh
Mehr anzeigen Weniger anzeigen
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.