Learn More
R&D Systems™ Recombinant Human TGF-beta 3 Protein
Beschreibung
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.01-0.04 ng/mL.
Spezifikation
Spezifikation
| Zusammensetzung | Lyophilisiert aus einer 0,2 μm gefilterten Lösung in Acetonitril und TFA. |
| Gen-ID (Entrez) | 7043 |
| Molekulargewicht | 12.7 kDa (monomer) |
| Name | TGF-beta 3 |
| Menge | 10 μg |
| Quelle | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TGF-beta 3 protein Ala301-Ser412 (Tyr340Phe) |
| Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Strukturform | Disulfide-linked homodimer |
| Gen-Alias | ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 |
| Konjugat | Unconjugated |
| Mehr anzeigen |