Learn More
R&D Systems™ Recombinant Human Thrombopoietin/TPO Protein, CF
Artikelnummer. 30306079
Click to view available options
Menge:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Artikelnummer. 30306079
Marke: R&D Systems™ BTTPO500
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Recombinant Protein
Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. Avanzi, G. et al. (1988) Br. J. Haematol. 69:359. The ED50 for this effect is 0.0500-0.500 ng/mL.
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer NP_000451.1
Zur Verwendung mit (Anwendung) Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gen-ID (Entrez) 7066
Molekulargewicht M.W.-theoretical: 19 kDa, M.W.-observed: 18-19 kDa, under reducing conditions.
Menge 500 μg
Quelle E. coli-derived human Thrombopoietin/Tpo protein Ser22-Leu195
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin-Konzentration <0.10 EU per 1 μg of the protein by the LAL method.
Gen-Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Konjugat Unconjugated
Rekonstitution Reconstitute the 10 μg size at 100 μg/mL in water. Reconstitute all other sizes at 500 μg/mL in water.
Protein Thrombopoietin/THPO