Learn More
Novus Biologicals™ Recombinant Human ZBTB7A/Pokemon GST (N-Term) Protein
Artikelnummer. 18602850 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18602850 10 μg 10 Mikrogramm
18614171 25 μg 25 Mikrogramm
2 options
Artikelnummer. 18602850
Marke: Novus Biologicals™ H00051341P0110ug
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Spezifikation
Zur Verwendung mit (Anwendung) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Zusammensetzung 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gen-ID (Entrez) 51341
Molekulargewicht MolecularWeight-theroretical: 37.62 kDa
Menge 10 μg
Lagerungsbedingungen Store at -80°C. Avoid freeze-thaw cycles.
Gen-Alias DKFZp547O146, Factor binding IST protein 1, Factor that binds to inducer of short transcripts protein 1, FBI-1POK erythroid myeloid ontogenic factor, FBI1TTF-I-interacting peptide 21, HIV-1 inducer of short transcripts binding protein, Leukemia/lymphoma-related factor, LRFHIV-1 1st-binding protein 1, lymphoma related factor, MGC99631, pokemon, TIP21, ZBTB7zinc finger and BTB domain containing 7, zinc finger and BTB domain containing 7A, zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcriptsbinding protein, zinc finger and BTB domain-containing protein 7A, Zinc finger protein 857A, ZNF857APOZ and Krueppel erythroid myeloid ontogenic factor
Gensymbol ZBTB7A
Forschungskategorie Cancer, Transcription Factors and Regulators
Reinheits- oder Qualitätsgrad >80% by SDS-PAGE and Coomassie blue staining
Protein ZBTB7A/Pokemon