missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human ZIC4 GST (N-Term) Protein
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Menge:
10 μg
25 μg
missing translation for 'unitSize'
10 Mikrogramm
25 Mikrogramm
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| Zur Verwendung mit (Anwendung) | Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification |
| Zusammensetzung | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gen-ID (Entrez) | 84107 |
| Molekulargewicht | MolecularWeight-theroretical: 35.64 kDa |
| Menge | 25 μg |
| Lagerungsbedingungen | Store at -80°C. Avoid freeze/thaw cycles. |
| Gen-Alias | FLJ42609, FLJ45833, Zic family member 4, Zinc finger protein of the cerebellum 4 zinc family member 4 protein HZIC4, zinc finger protein ZIC 4 |
| Gensymbol | ZIC4 |
| Reinheits- oder Qualitätsgrad | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | ZIC4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction