missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Carboxylesterase 1C Protein

Artikelnummer. 16087620
Click to view available options
Menge:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
200 Mikrogramm
50 Mikrogramm
500 Mikrogramm
This item is not returnable. View return policy

Product Code. 16087620

Brand: enQuireBio™ QP7188ye500ug

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Carboxylesterase 1C, was expressed in the hosts and tags indicated.

Specifications

Zugriffsnummer NP_031980.2
Gen-ID (Entrez) 13884
Name Carboxylesterase 1C Protein
Menge 500 μg
Kennzeichnung Research Use Only
Endotoxin-Konzentration Not determined.
Gensymbol Ces1c
Proteinlänge Partial (see sequence information for more details).
Produkttyp Recombinant Protein
Kreuzreaktivität Mouse
Spezies Yeast
Proteinmarkierung His
Sequenz HSLLPPVVDT TQGKVLGKYI SLEGFEQPVA VFLGVPFAKP PLGSLRFAPP QPAEPWSFVK NATSYPPMCS QDAGWAKILS DMFSTEKEIL PLKISEDCLY LNIYSPADLT KSSQLPVMVW IHGGGLVIGG ASPYNGLALS AHENVVVVTI QYRLGIWGLF STGDEHSPGN WAHLDQLAAL RWVQDNIANF GGNPDSVTIF GESSGGISVS VLVLSPLGKD LFHRAISESG VVINTNVGKK NIQAVNEIIA TLSQCNDTSS AAMVQCLRQK TESELLEISG KLVQYNISLS TMIDGVVLPK APEEILAEKS FNTVPYIVGF NKQEFGWIIP MMLQNLLPEG KMNEETASLL LRRFHSELNI SESMIPAVIE QYLRGVDDPA KKSELILDMF GDIFFGIPAV LLSRSLRDAG VSTYMYEFRY RPSFVSDKRP QTVEGDHGDE IFFVFGAPLL KEGASEEETN LSKMVMKFWA NFARNGNPNG EGLPHWPEYD EQEGYLQIGA TTQQAQRLKA EEVAFWTELL AKNPPETDPT EH
Puffer Tris-based buffer, 50% glycerol
Reinheits- oder Qualitätsgrad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.