Learn More
enQuireBio™ Recombinant Mouse IL-11 / interleukin 11 Protein
Artikelnummer. 15996199
Ansicht ändern
Click to view available options
Menge:
1 mg
500 μg
Packungsgröße:
1 Milligramm
500 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15996199 500 μg 500 Mikrogramm
15986199 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15996199 Lieferant enQuireBio™ Lieferanten-Nr. QP5472500ug
Please to purchase this item. Need a web account? Register with us today!
Recombinant Protein
Spezifikation
Zugriffsnummer NP_032376.1
Gen-ID (Entrez) 16156
Name IL-11 / interleukin 11 Protein
Menge 500 μg
Kennzeichnung Research Use Only
Endotoxin-Konzentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gensymbol IL11
Biologische Aktivität Shows bioactivity at a concentration of 2.5 ng/mL in the T11 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^5 units/mg.
Produkttyp Recombinant Protein
Kreuzreaktivität Mouse
Spezies E. coli
Proteinmarkierung Untagged
Sequenz MPGPPAGSPR VSSDPRADLD SAVLLTRSLL ADTRQLAAQM RDKFPADGDH SLDSLPTLAM SAGTLGSLQL PGVLTRLRVD LMSYLRHVQW LRRAGGPSLK TLEPELGALQ ARLERLLRRL QLLMSRLALP QAAPDQPVIP LGPPASAWGS IRAAHAILGG LHLTLDWAVR GLLLLKTRL
Puffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Reinheits- oder Qualitätsgrad Greater than 95% as determined by SDS-PAGE.