Learn More
enQuireBio™ Recombinant Mouse Kallikrein-14 Protein
Artikelnummer. 16068811
Click to view available options
Menge:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
200 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Artikelnummer. 16068811
Marke: enQuireBio™ QP9831ye100ug
om dit product te kopen Registreer vandaag om een webaccount aan te maken
A DNA sequence encoding the Mus musculus (Mouse) Kallikrein-14, was expressed in the hosts and tags indicated.
Spezifikation
Zugriffsnummer NP_777355.1
Gen-ID (Entrez) 317653
Name Kallikrein-14 Protein
Menge 100 μg
Kennzeichnung Research Use Only
Endotoxin-Konzentration Not determined.
Gensymbol Klk14
Proteinlänge Full. Length. of. Mature. Protein
Produkttyp Recombinant Protein
Kreuzreaktivität Mouse
Spezies Yeast
Proteinmarkierung His
Sequenz IIGGYRCVRN SQPWQVALQA GPGHRFLCGG VLLSDQWVIT AAHCARPILH VALGKHNIRR WEATQQVVRV ARQVPHPQYQ PQAHDNDLML LKLQKKVRLG RAVKTISVAS SCASPGTPCR VSGWGTIASP IARYPTALQC VNVNIMSEQA CHRAYPGIIT SGMVCAGVPE GGKDSCQGDS GGPLVCGGQL QGLVSWGMER CAMPGYPGVY ANLCNYHSWI QRTMQSN
Puffer Tris-based buffer, 50% glycerol
Reinheits- oder Qualitätsgrad Greater than 84% as determined by SDS-PAGE.