Learn More
R&D Systems™ Recombinant Mouse LIGHT/TNFSF14 Protein
Artikelnummer. 18126630
Click to view available options
Zusammensetzung:
Lyophilisiert aus einer 0,2 μm gefilterten Lösung in PBS mit BSA als Trägerprotein
Lyophilisiert aus einer 0,2 μm gefilterten Lösung in PBS.
Packungsgröße:
25 Mikrogramm
Artikelnummer. 18126630
Marke: R&D Systems™ 1794LT025
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Measured in a cytotoxicity assay using HT-29 human colon adenocarcinoma cells Yu, K.Y. et al. (1999) J. Biol. Chem. 274:13733; Harrop, J.A. et al. (1998) J. Biol. Chem. 273:27548; Zhai, Y. et al. (1998) J. Clin. Invest. 102:1142. The ED50 for this effect is 2-5 μg/mL in the presence of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ) and recombinant human IFN-gamma .
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer Q9QYH9
Zur Verwendung mit (Anwendung) Bioactivity
Zusammensetzung Lyophilisiert aus einer 0,2 μm gefilterten Lösung in PBS mit BSA als Trägerprotein
Gen-ID (Entrez) 8740
Molekulargewicht M.W. (Observed): 24-34 kDa, reducing conditions; M.W. (theoretical): 21 kDa
Name LIGHT/TNFSF14
Menge 25 μg
Quelle Mouse myeloma cell line, NS0-derived mouse LIGHT/TNFSF14 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse LIGHT (Asp72-Val239) (N-terminus) Accession # Q9QYH9 (C-terminus)
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin-Konzentration <0.10 EU / 1 μg of the protein by the LAL method.
Gensymbol Tnfsf14
Konjugat Unconjugated
Rekombinant Recombinant
Rekonstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Reinheits- oder Qualitätsgrad >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LIGHT/TNFSF14