Learn More
R&D Systems™ Recombinant Mouse TNF-alpha (aa 80-235) Protein
Artikelnummer. p-7145981
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18717666 25 μg 25 Mikrogramm
18728496 100 μg 100 Mikrogramm
2 options
Artikelnummer. 18717666
Marke: R&D Systems™ 410MT025
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Mouse TNF-alpha (aa 80-235) Protein is derived from E. coli. The Recombinant Mouse TNF-alpha (aa 80-235) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer P06804
Zur Verwendung mit (Anwendung) Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gen-ID (Entrez) 7124
Molekulargewicht 17 kDa
Menge 25 μg
Quelle <EM>E. coli</EM>-derived mouse TNF-alpha protein Leu80-Leu235, with an N-terminal Met
Lagerungsbedingungen Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Kennzeichnung RUO
Endotoxin-Konzentration <0.01 EU per 1 μg of the protein by the LAL method.
Gen-Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gensymbol Tnf
Konjugat Unconjugated
Kreuzreaktivität TNF-alpha
Spezies Mouse
Rekonstitution Reconstitute at 100 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Reinheits- oder Qualitätsgrad >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.