missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse VEGF 164 Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
10 μg
100 μg
1000 μg
500 μg
Packungsgröße:
10 Mikrogramm
100 Mikrogramm
1000 Mikrogramm
500 Mikrogramm
Beschreibung
Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human umbilical vein endothelial cells(HUVEC) is between 1.0-5.0 ng/mL.
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Bioactivity |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gen-ID (Entrez) | 7422 |
| Molekulargewicht | MolecularWeight-theroretical: 40 kDa |
| Menge | 10 μg |
| Lagerungsbedingungen | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin-Konzentration | Less than 0.01 EU/μg of VEGF 164 AS determined by LAL method. |
| Gensymbol | VEGFA |
| Rekonstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Inhibitor-Eigenschaften | As a result of glycosylation, it migrates to at least two bands with molecular weights ranging from approximately 40 kDa in SDS-PAGE under non-reducing conditions. |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur