Learn More
Novus Biologicals™ Recombinant Rat VEGF 164 Protein
Artikelnummer. 18699915 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
10 μg
100 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
18699915 100 μg 100 Mikrogramm
18620386 10 μg 10 Mikrogramm
18670386 1 mg 1 Milligramm
18694765 500 μg 500 Mikrogramm
4 options
Artikelnummer. 18699915
Marke: Novus Biologicals™ NBP235310100ug
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Protein
VEGF 165 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human umbilical vein endothelial cells(HUVEC) is less than 5 ng/mL, corresponding to a specific activity of > 2.0 x 10^5 IU/mg.
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer P16612
Zur Verwendung mit (Anwendung) SDS-PAGE, Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM Tris, 300 mM NaCl, pH 8.8.
Gen-ID (Entrez) 7422
Molekulargewicht MolecularWeight-theroretical: 38.7 kDa
Menge 100 μg
Lagerungsbedingungen Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin-Konzentration Less than 1 EU/μg of VEGF 164 AS determined by LAL method.
Gensymbol VEGFA
Rekonstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Forschungskategorie Biologically Active Proteins
Reinheits- oder Qualitätsgrad >95%, by SDS-PAGE and HPLC
Protein VEGF 164