missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rhesus Macaque S100B Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
10 μg
100 μg
1000 μg
500 μg
Packungsgröße:
10 Mikrogramm
100 Mikrogramm
1000 Mikrogramm
500 Mikrogramm
Specifications
Specifications
| Zur Verwendung mit (Anwendung) | SDS-PAGE |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4. |
| Gen-ID (Entrez) | 6285 |
| Molekulargewicht | MolecularWeight-theroretical: 10.6 kDa |
| Menge | 500 μg |
| Lagerungsbedingungen | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin-Konzentration | Less than 1 EU/μg of S100B as determined by LAL method. |
| Gen-Alias | beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10 protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta |
| Gensymbol | S100B |
| Rekonstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction