missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15684-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.Spezifikation
| REEP2 | |
| Polyclonal | |
| Unconjugated | |
| REEP2 | |
| C5orf19, receptor accessory protein 2, REEP2, SGC32445 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225362, 51308, 682105 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VCD6, Q9BRK0 | |
| REEP2 | |
| REEP2 Fusion Protein Ag8186 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur