Learn More
Thermo Scientific™ Remel™ PathoDx™ Respiratory Virus Panel Kontrollobjektträger
Artikelnummer. 11414139
Ansicht ändern
Click to view available options
Menge:
5 Objektträger/Pk.
Packungsgröße:
5 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
11414139 5 Objektträger/Pk. 5 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11414139 Lieferant Thermo Scientific™ Lieferanten-Nr. R62414
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Der Remel PathoDx Respiratory Virus Panel Kontrollobjektträger ist zur Verwendung mit dem PathoDx Respiratory Virus Panel vorgesehen.
The Negative Control Reagent contains fluorescein-labeled murine antibodies which do not react with the viral agents.
  • Ready to use - Prepared reagents
  • Easy to store - Store at 2-8°C
  • Easy to interpret - Uninfected cells will stain red due to the Evans blue counterstain in each staining reagent
Negative Control Reagent (Red Cap) is one dropper bottle containing 10.0 mL of fluorescein-labeled purified murine IgG antibodies that are non-reactive with any respiratory virus, and Evans blue counterstain in a protein-stabilized buffer solution, with 0.098% sodium azide as preservative.
Each control slide has eight wells and each well is clearly marked with the type of virus-infected cell that is on the slide. One of the wells contains uninfected LLC-MK2 cells and the slide is fixed in acetone.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Spezifikation
Beschreibung PathoDx™Kontroll-Objektträger für Atmungsvirus-Panel
Typ Regelung
Menge 5 Objektträger/Pk.