Learn More
Restek™ Injector/Detector Plug Nut
Beschreibung
- Use to cap off an injector to isolate leaks
- Use to cap off a detector for thermal cleaning
- Use to check a detector or make-up gas flow rate
- Use to cap off a detector and prevent hydrogen from accidentally diffusing into the oven from an unused detector base
Spezifikation
Spezifikation
| Typ | Mutter |
| Zur Verwendung mit (Anwendung) | Cap Off or Check Injector or Detector for Leaks, Cleaning, Flow-Rate. |
| Verpackungsmenge | 2/Karton |
| Zur Verwendung mit | HP GC |