missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFWD2 Recombinant Rabbit Monoclonal Antibody (5A0S9)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Spezifikation
| Antigen | RFWD2 |
|---|---|
| Klon | 5A0S9 |
| Konzentration | 1.16 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Western Blot, Immunocytochemistry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30283344
|
Invitrogen™
MA557925 |
20 μL |
176.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
30284627
![]() |
Invitrogen™
MA557926 |
100 μL |
436.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Immunogen sequence: YYDLRNTKQP IMVFKGHRKA VSYAKFVSGE EIVSASTDSQ LKLWNVGKPY CLRSFKGHIN EKNFVGLASN GDYIACGSEN NSLYLYYKGL SKTLLTFKFD TVKSVLDKDR KEDDTNEFVS AVCWRALPDG ESNVLIAANS QGTIKVLELV.
E3 ubiquitin-protein ligase that mediates ubiquitination and subsequent proteasomal degradation of target proteins. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Involved in JUN ubiquitination and degradation. Directly involved in p53 (TP53) ubiquitination and degradation, thereby abolishing p53-dependent transcription and apoptosis. Ubiquitinates p53 independently of MDM2 or RCHY1. Probably mediates E3 ubiquitin ligase activity by functioning as the essential RING domain subunit of larger E3 complexes. In contrast, it does not constitute the catalytic RING subunit in the DCX DET1-COP1 complex that negatively regulates JUN, the ubiquitin ligase activity being mediated by RBX1. Involved in 14-3-3 protein sigma/SFN ubiquitination and proteasomal degradation, leading to AKT activation and promotion of cell survival. Ubiquitinates MTA1 leading to its proteasomal degradation. Upon binding to TRIB1, ubiquitinates CEBPA, which lacks a canonical COP1-binding motif (Probable). [UniProt]Spezifikation
| RFWD2 | |
| 1.16 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9R1A8 | |
| 26374, 360860 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 582-731 of human COP1/RFWD2 (NP_071902.2). | |
| Antibody |
| 5A0S9 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| RFWD2 | |
| AI316802; C80879; Constitutive photomorphogenesis protein 1 homolog; constitutive photomorphogenic protein 1; COP1; COP1D; E3 ubiquitin-protein ligase COP1; E3 ubiquitin-protein ligase RFWD2; hCOP1; mCOP1; putative ubiquitin ligase COP1; Rfwd2; RGD1304773; ring finger and WD repeat domain 2; ring finger and WD repeat domain 2, E3 ubiquitin protein ligase; RING finger and WD repeat domain protein 2; RING finger protein 200; RING-type E3 ubiquitin transferase RFWD2; RNF200 | |
| RFWD2 | |
| Primary | |
| Protein A |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts