Learn More
Merck Rheodyne™ Injector, Rheodyne™, 7725i
Internal position sensing switch, Manual, Number of ports: 6 and Number of positions: 2
Marke: Merck 57621
Weitere Details : Netto-Gewicht : 0.20000kg
Beschreibung
Rheodyne™ Injector model is a stainless steel version and comprises of a six-port sample injection valves in which the sample is loaded through a built-in needle port in the front of the valve.Spezifikation
Liquid Chromatography | |
7,000 psi max. pressure |
Rheodyne is a registered trademark of IDEX Health & Science, LLC