Learn More
RHOF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. 16899913 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16899913 20 μL 20 Mikroliter
16889913 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16899913 Lieferant Proteintech Lieferanten-Nr. 122901AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
Plasma membrane-associated small GTPase which cycles between an active GTP-bound and an inactive GDP-bound state. Causes the formation of thin, actin-rich surface projections called filopodia. Functions cooperatively with CDC42 and Rac to generate additional structures, increasing the diversity of actin-based morphology.
Spezifikation
Antigen RHOF
Anwendungen Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 0.13 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen RHOF
Gen-Zugriffsnummer Q8BYP3, Q9HBH0
Gen-Alias ARHF, Rho family GTPase Rif, Rho in filopodia, RHOF, RIF
Gensymbole RHOF
Wirtsspezies Rabbit
Immunogen RHOF Fusion Protein Ag2943
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 23912, 54509, 690130
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG