missing translation for 'onlineSavingsMsg'
Learn More

RMI2 Antibody - BSA Free, Novus Biologicals™

Artikelnummer. 18605532 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.02 mL
0.1 mL
Packungsgröße:
0.01 Milliliter
0.02 Milliliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18605532 0.02 mL 0.02 Milliliter
18605182 0.1 mL 0.01 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18605532 Lieferant Novus Biologicals Lieferanten-Nr. NBP2947390.02ml

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

RMI2 Polyclonal antibody specifically detects RMI2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen RMI2
Anwendungen Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Western Blot 1:500-1:2000
Zusammensetzung PBS (pH 7.3), 50% glycerol
Gen-Alias BLAP18RMI2, BLM-associated protein of 18 kDa, chromosome 16 open reading frame 75, hRMI2, MGC24665, RecQ-mediated genome instability 2, S. cerevisiae, homolog of, recQ-mediated genome instability protein 2
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human RMI2 (NP_689521.1). MAAAADSFSGGPAGVRLPRSPPLKVLAEQLRRDAEGGPGAWRLSRAAAGRGPLDLAAVWMQGRVVMADRGEARLRDPSGDFSVRGLERVPRGRPCLVPGKYVMVMGVVQACSPEPCLQAVKMTDLSDNPIHESMWELEVEDLHRNIP
Reinigungsverfahren Affinity purified
Menge 0.02 mL
Regulatorischer Status RUO
Forschungsgebiet Direct Reversal of DNA Damage
Primär oder sekundär Primary
Gen-ID (Entrez) 116028
Zielspezies Human
Inhalt und Lagerung Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.