missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE7 Antibody (CL0224) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-43953
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
RNASE7 Monoclonal antibody specifically detects RNASE7 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Spezifikation
| RNASE7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 3.1.27, EC 3.1.27.-, EC 3.1.27.5, MGC133220, ribonuclease 7, ribonuclease, RNase A family, 7, RNase 7, SAP-2, Skin-derived antimicrobial protein 2 | |
| This antibody was generated using a recombinant protein sequence of Q9H1E1, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 84659 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL0224 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur