Learn More
Tocris Bioscience™ Ro 25-6981 maleate
Click to view available options
Menge:
1 mg
10 mg
50 mg
Packungsgröße:
1 Milligramm
10 Milligramm
50 Milligramm
Beschreibung
Ro 25-6981 maleate is a potent and selective activity-dependent blocker of NMDA receptors containing the GluN2B (formally NR2B) subunit. IC50 values are 0.009 and 52 μM for cloned receptor subunit combinations GluN1C/GluN2B and GluN1C/GluN2A respectively. Displays neuroprotectant effects in vivo and in vitro.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Spezifikation
Spezifikation
| Chemischer Name oder Material | (αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate |
| CAS | 1312991-76-6 |
| Prozentgehaltsbereich | >98% |
| Menge | 50 mg |
| Löslichkeit | Soluble to 100mM in DMSO |
| Summenformel | C22H29NO2.C4H4O4 |
| Formelmasse | Observed MW: 455.55 |
Nur für Forschungszwecke