Beschreibung
The ROR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ROR1. This antibody recognizes human antigen. The ROR1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Spezifikation
Spezifikation
| Antigen | ROR1 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA |
| Klassifikation | Monoclonal |
| Klon | 1F5D8 |
| Konzentration | 1000 μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Menge | 20 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 4919 |
| Mehr anzeigen |