Learn More
Roxatidinacetathydrochlorid, TRC
Specifications
Specifications
| Chemischer Name oder Material | Roxatidine Acetate Hydrochloride |
| CAS | 93793-83-0 |
| Summenformel | C19 H28 N2 O4 . Cl H |
| Menge | 10 g |
| Synonym | Acetamide','2-(acetyloxy)-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]-','hydrochloride (1:1), Acetamide','2-(acetyloxy)-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]-','monohydrochloride (9CI), Altat, Gastralgin, HOE 760, Neo H2, Roxatidine acetate hydrochloride, Roxit, TZU 0460, Zarocs |
| InChI-Formel | InChI=1S/C19H28N2O4.ClH/c1-16(22)25-15-19(23)20-9-6-12-24-18-8-5-7-17(13-18)14-21-10-3-2-4-11-21;/h5,7-8,13H,2-4,6,9-12,14-15H2,1H3,(H,20,23);1H |
| SMILES | Cl.CC(=O)OCC(=O)NCCCOc1cccc(CN2CCCCC2)c1 |
| IUPAC-Name | [2-Oxo-2-[3-[3-(Piperidin-1-ylmethyl)phenoxy]propylamino]ethyl]acetat;Hydrochlorid |
| Molekulargewicht (g/mol) | 384.9 |
| Formelmasse | 384.1816 |