missing translation for 'onlineSavingsMsg'
Learn More

RP11-195F19.5 Antibody, R&D Systems™

Artikelnummer. 18601839 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
25 μL
0.1 mL
Packungsgröße:
0.10 Milliliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18601839 25 μL 25 Mikroliter
18649458 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18601839 Lieferant Novus Biologicals Lieferanten-Nr. NBP24960925ul
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

RP11-195F19.5 Polyclonal specifically detects RP11-195F19.5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen RP11-195F19.5
Anwendungen Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Zusammensetzung PBS (pH 7.2), 40% Glycerol
Gen-Alias CDNA FLJ50015, HCG2040265, Isoform CRA_a, LOC730098, Novel Protein, Uncharacterized protein, Uncharacterized Protein 13
Wirtsspezies Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: PLQETFGAQDMSGMRRYEVALEAEEEIYWGCFYFFPWLRMWRRERSSAHPGEQKLEPLRGLMSCLSSGLGPTPQRSGRGFPRRSPTAAAQPASALK
Reinigungsverfahren Immunogen affinity purified
Menge 25 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Produkttyp Antibody
Form Purified
Isotype IgG
Produktlinie Novus
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.