missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
Spezifikation
Spezifikation
| Antigen | RPL7 |
| Anwendungen | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Klassifikation | Polyclonal |
| Konzentration | 0.45 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | RPL7 |
| Gen-Zugriffsnummer | P05426, P14148, P18124 |
| Gen-Alias | 60S ribosomal protein L7, humL7 1, ribosomal protein L7, RPL7 |
| Gensymbole | RPL7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?