Learn More
Tocris Bioscience™ (RS)-3,5-DHPG
Click to view available options
Menge:
1 mg
10 mg
50 mg
Packungsgröße:
1 Milligramm
10 Milligramm
50 Milligramm
Beschreibung
(RS)-3,5-DHPG is a selective group I metabotropic glutamate receptor agonist which activates both mGlu1 and mGlu5. Also reported to be an antagonist at metabotropic glutamate receptors linked to phospholipase D.
S-enantiomer also available.
Spezifikation
Spezifikation
| Chemischer Name oder Material | (RS)-3,5-Dihydroxyphenylglycine |
| CAS | 19641-83-9 |
| Prozentgehaltsbereich | >99% |
| Menge | 1 mg |
| Löslichkeit | Soluble to 10mM in water |
| Summenformel | C8H9NO4 |
| Formelmasse | Observed MW: 183.16 |
Nur für Forschungszwecke