missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RTN4RL1 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 30535533
Change view
Click to view available options
Menge:
100 μg
Artikelnummer. Menge
30535533 100 μg
1 options
Dieser Artikel kann nicht zurückgegeben werden. View return policy

Product Code. 30535533

Brand: Invitrogen™ PA5148642

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

RTN4RL1 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN4RL1
Anwendungen Western Blot
Klassifikation Polyclonal
Konzentration 0.2 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with trehalose and no preservative
Gen Rtn4rl1
Gen-Zugriffsnummer Q86UN2
Gen-Alias NgR3; Ngrh2; NGRL2; Nogo receptor-like 2; Nogo-66 receptor homolog 2; Nogo-66 receptor-related protein 3; reticulon 4 receptor like 1; reticulon 4 receptor-like 1; reticulon-4 receptor-like 1; RGD:727812}; Rtn4rl1; rtn4rl1 {ECO:0000312
Gensymbole Rtn4rl1
Wirtsspezies Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human NgR3/NgRH2, Cys25-Ala419.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 146760
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.