Learn More
(S)-(+)-N-Carboxybenzoyl-4-Amino-2-hydroxybuttersäure, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | (S)-(+)-N-Carboxybenzoyl-4-Amino-2-hydroxybutyric Acid |
| CAS | 40371-50-4 |
| Summenformel | C12H15NO5 |
| Menge | 5 g |
| Synonym | (S)-2-Hydroxy-4-[[(phenylmethoxy)carbonyl]amino]butyric Acid, (S)-4-(Benzyloxycarbonylamino)-2-hydroxybutyric Acid, (S)-4-[(Benzyloxycarbonyl)amino]-2-hydroxybutanoic Acid, L(-)-gamma-Benzyloxycarbonylamino-alpha-hydroxybutyric Acid, L-gamma-Benzyloxycarbonylamino-alpha-hydroxybutyric Acid |
| InChI-Formel | InChI=1S/C12H15NO5/c14-10(11(15)16)6-7-13-12(17)18-8-9-4-2-1-3-5-9/h1-5,10,14H,6-8H2,(H,13,17)(H,15,16)/t10-/m0/s1 |
| SMILES | O[C@@H](CCNC(=O)OCc1ccccc1)C(=O)O |
| IUPAC-Name | (2S)-2-Hydroxy-4-(phenylmethoxycarbonylamino)butansäure |
| Molekulargewicht (g/mol) | 253.25 |
| Formelmasse | 253.095 |