Learn More
S100 calcium binding protein A14 Antibody (S100A14/9077R), Alexa Fluor™ 750, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
S100 calcium binding protein A14 Monoclonal antibody specifically detects S100 calcium binding protein A14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | S100 calcium binding protein A14 |
| Anwendungen | ELISA, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | S100A14/9077R |
| Konjugat | Alexa Fluor 750 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant full-length human S100 calcium binding protein A14 protein |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |