Learn More
Novus Biologicals S100B Antibody (4C4.9), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-33168APC
Product Code. 18439281
  • ONLINE EXCLUSIVE PRICE
    647.00 €
    611.10 € / 0.10 Milliliter
    Sparen 35.90 €
    5% Rabatt
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
This item is not returnable. View return policy
Description
Description
S100B Monoclonal antibody specifically detects S100B in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S100B
Monoclonal
APC
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Purified bovine brain S100B protein (Uniprot: P04271)
0.1 mL
Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells
6285
Store at 4°C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
4C4.9
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat, Bovine
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications