missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
S1P3/EDG-3 Monoclonal antibody specifically detects S1P3/EDG-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Spezifikation
Spezifikation
| Antigen | S1P3/EDG-3 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | SR1706 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Zusammensetzung | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gen-Alias | EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 |
| Wirtsspezies | Rabbit |
| Immunogen | A synthesized peptide derived from human S1P3/EDG-3 (Uniprot #: Q99500) |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?