Learn More
Invitrogen™ SALF Monoclonal Antibody (1F3)
GREENER_CHOICE
Artikelnummer. 15538972
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
15538972 100 μg 100 Mikrogramm
1 options
Artikelnummer. 15538972
Marke: Invitrogen™ MA521231
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
Peptide Sequence: SLKSVVVVQG AYVELQAFVN MASLAQRSSY AGSLRSCDNI RIHFPVPSQW IKALWTMNLQ RQKSLKAKMN RRACLGSLQE LESEPVIQVT VG.
Endocytosis of cell surface proteins is mediated by a complex molecular machinery that assembles on the inner surface of the plasma membrane. This gene encodes one of two human homologs of the Drosophila melanogaster stoned B protein. This protein is related to components of the endocytic machinery and exhibits a modular structure consisting of an N-terminal proline-rich domain, a central region of homology specific to the human stoned B-like proteins, and a C-terminal region homologous to the mu subunits of adaptor protein (AP) complexes. Co-transcription of this gene and the neighboring downstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen SALF
Anwendungen ELISA, Immunocytochemistry
Klassifikation Monoclonal
Klon 1F3
Konzentration 0.2-1.0 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative; pH 7.4
Gen STON1
Gen-Zugriffsnummer Q9Y6Q2
Gen-Alias 4921524J06Rik; C86231; Salf; Sblf; STN1; STNB1; STON1; STON1-GTF2A1L; STON1-GTF2A1L protein; STON1-GTF2A1L readthrough; stoned B homolog 1; stoned B/TFIIA-alpha/beta-like factor; stoned B-like factor; stoned-b1; stonin 1; Stonin1; stonin-1
Gensymbole STON1
Wirtsspezies Mouse
Immunogen STON1 (NP_006864, 529 a.a. ~ 620 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Reinigungsverfahren Affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 11037
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG2a κ