Learn More
Salmonella Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Marke:  Novus Biologicals NB600-1087
Artikelnummer. 18027673
  • ONLINE EXCLUSIVE PRICE
    360.00 €
    340.20 € / 0.25 Milliliter
    Sparen 19.80 €
    5% Rabatt
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Salmonella Polyclonal specifically detects Salmonella in Mouse, Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Salmonella
Polyclonal
Unconjugated
0.01 M Phosphate Buffered Saline, pH 7.2 with 0.1% Sodium Azide
Mixture of S. enteriditis, S. typhimurium, and S. heidelburg
0.25 mL
Primary
Bacteria
Purified
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
4.5 mg/mL
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Rabbit
Protein A purified
RUO
This is polyvalent for Salmonella “O” & “H” antigens. Immunocaptures Salmonellae. Antiserum is not absorbed and does react with related Enterobacteriaceae.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen