missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB104742-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Nucleocapsid Monoclonal antibody specifically detects Nucleocapsid in SARS-CoV-2 samples. It is validated for Western Blot, Western Blot, ELISA
Spezifikation
| Nucleocapsid | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 1035143 | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, ELISA | |
| YP_009724397.2 | |
| Spodoptera frugiperda, Sf 21-derived SARS-CoV-2 Nucleocapsid, Met1-Ala419, Accession # YP_009724397.2 | |
| 25 μg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur