missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences SARS-CoV-2 Peptide ILLNKHIDA (HLA-A*0201)
Alle ansehen IBA Lifesciences Produkte

Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
Description
- SARS-CoV-2 peptide ILLNKHIDA (HLA-A*0201) is a single peptide for stimulation of T cells.
- The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Specifications
| Reinheit | >85% |
| Endotoxin-Gehalt | Free |
| Format | Lyophilisiertes Pulver |
| Menge | 1 mg |
| Forschungskategorie | T cell stimulation |
Nur für Forschungszwecke.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction