missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV/SARS-CoV-2 ORF7a Monoclonal Antibody (3C9)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA535944
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SARS-CoV/SARS-CoV-2 ORF7a Monoclonal Antibody for Western Blot, ICC/IF
ORF7a is a SARS-CoV accessory protein that is composed of a type I transmembrane protein that localizes primarily to the Golgi apparatus. It can also be found on the cell surface. Its exact role in pathogenesis and disease is unknown. However, it has been observed to bind to BST-2 and inhibit BST-2 glycosylation, resulting in the subsequent loss of BST-2 antiviral function.
Spezifikation
| SARS-CoV/SARS-CoV-2 ORF7a | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| 0 | |
| ORF7a | |
| SARS-CoV ORF7a protein (16-111 a.a.). | |
| 100 μL | |
| Primary | |
| Virus | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 3C9 | |
| Unconjugated | |
| ORF7a | |
| 2019-nCoV orf7a; Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 orf7a; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS-CoV orf7a; SARS-CoV-2; SARS-CoV-2 orf7a | |
| Mouse | |
| Protein G | |
| RUO | |
| 43740573 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur