Learn More
Invitrogen™ SARS/SARS-CoV-2 Nucleocapsid Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA581794
Beschreibung
PA5-81794 cross reacts with SARS-CoV-2 (COVID-19) Nucleoprotein in ELISA and WB. This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SARS Coronavirus Nucleoprotein/NP.
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
Spezifikation
| SARS/SARS-CoV-2 Nucleocapsid | |
| Polyclonal | |
| Unconjugated | |
| N | |
| COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 43740575 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P0DTC9 | |
| N | |
| Recombinant Human SARS Coronavirus Nucleoprotein/NP Protein. | |
| 100 μL | |
| Primary | |
| Virus | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.