missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCAP Recombinant Rabbit Monoclonal Antibody (HL2796)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556869
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SCAP Recombinant Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. This protein contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced transcript variants that encode distinct isoforms have been identified.
Spezifikation
| SCAP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| A2RRU4, Q12770, Q6GQT6 | |
| SCAP | |
| Recombinant protein of human SCAP. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2796 | |
| Unconjugated | |
| SCAP | |
| 9530044G19; EMBL:AAI31853.1}; KIAA0199; mKIAA0199; PSEC0227; SCAP; scap {ECO:0000312; Sreb cleavage-activating protein; SREBF chaperone; SREBP cleavage activating protein; SREBP cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; sterol regulatory element-binding protein cleavage-activating protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22937, 235623, 301024 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur