missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
SCP1 Polyclonal specifically detects SCP1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.
Spezifikation
Spezifikation
| Antigen | SCP1 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot |
| Klassifikation | Polyclonal |
| Konjugat | DyLight 488 |
| Verdünnung | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting |
| Zusammensetzung | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gen-Zugriffsnummer | Q15431 |
| Gen-Alias | Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 |
| Gensymbole | SYCP1 |
| Wirtsspezies | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?