missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDHC Recombinant Rabbit Monoclonal Antibody (24GB1075)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557706
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SDHC Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes one of four nuclear-encoded subunits that comprise succinate dehydrogenase, also known as mitochondrial complex II, a key enzyme complex of the tricarboxylic acid cycle and aerobic respiratory chains of mitochondria. The encoded protein is one of two integral membrane proteins that anchor other subunits of the complex, which form the catalytic core, to the inner mitochondrial membrane. There are several related pseudogenes for this gene on different chromosomes. Mutations in this gene have been associated with paragangliomas. Alternatively spliced transcript variants have been described.
Spezifikation
| SDHC | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SDHC | |
| 0610010E03Rik; AI316496; AU019277; CYB560; CYBL; cytochrome B large subunit of complex II; integral membrane protein CII-3; integral membrane protein CII-3b; large subunit of cytochrome b; PGL3; QPs1; QPs-1; SDH3; SDHC; succinate dehydrgenase cytochrome b; succinate dehydrogenase 3, integral membrane subunit; succinate dehydrogenase complex subunit C; succinate dehydrogenase complex subunit C integral membrane protein 15kDa; succinate dehydrogenase complex, subunit C, integral membrane protein; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa; succinate dehydrogenase cytochrome b560 subunit, mitochondrial; Succinate-ubiquinone oxidoreductase cytochrome B large subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 289217, 6391, 66052 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB1075 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99643, Q9CZB0 | |
| SDHC | |
| A synthesized peptide derived from human SDHC (aa 10-69). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur