Learn More
SDSL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
391.65 € - 509.25 €
Spezifikation
Antigen SDSL
Verdünnung Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20
Klassifikation Polyclonal
Konjugat Unconjugated
Wirtsspezies Rabbit
Mehr Spezifikationen ansehen
Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
18423971
Unterlagen ansehen
Novus Biologicals
NBP1-87058-25ul
25 μL
ONLINE EXCLUSIVE PRICE
415.00 €
391.65 € / 25 Mikroliter
Sparen 23.35 €
5% Rabatt
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
18211836
Unterlagen ansehen
Novus Biologicals
NBP1-87058
0.1 mL
ONLINE EXCLUSIVE PRICE
539.00 €
509.25 € / 0.10 Milliliter
Sparen 29.75 €
5% Rabatt
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
SDSL Polyclonal specifically detects SDSL in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
Spezifikation
SDSL
Polyclonal
Rabbit
metabolism
EC 4.3.1.17, EC 4.3.1.19, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDH, SDH 2, SDS-RS1, Serine dehydratase 2, serine dehydratase related sequence 1, serine dehydratase-like, TDH
SDSL
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
113675
This antibody was developed against Recombinant Protein corresponding to amino acids:RHLVCSSGGNAGIAAAYAARKLGIPATIVLPESTSLQVVQRLQGEGAEVQLTGKVWDEANLRAQELAKRDGWENVPPFDHPLIWKGHASLVQELKAVLRT
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.