missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDSL Monoclonal Antibody (OTI1A8)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA526075
This item is not returnable.
View return policy
Description
SDSL Monoclonal Antibody for Western Blot, IHC (P), Flow
SDSL has low serine dehydratase and threonine dehydratase activity.
Specifications
| SDSL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96GA7 | |
| SDSL | |
| Full length human recombinant protein of SDSL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A8 | |
| Unconjugated | |
| SDSL | |
| 4432411H13Rik; AI504310; cSDH; L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; SDH1; Sds; SDSL; SDS-RS1; Serine dehydratase 2; serine dehydratase like; serine dehydratase related sequence 1; serine dehydratase-like; TDH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 113675 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?