Learn More
Invitrogen™ Securin Recombinant Rabbit Monoclonal Antibody (24GB1495)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557654
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Securin Recombinant Monoclonal Antibody for Western Blot, Flow
The encoded protein is a homolog of yeast securin proteins, which prevent separins from promoting sister chromatid separation. It is an anaphase-promoting complex (APC) substrate that associates with a separin until activation of the APC. The gene product has transforming activity in vitro and tumorigenic activity in vivo, and the gene is highly expressed in various tumors. The gene product contains 2 PXXP motifs, which are required for its transforming and tumorigenic activities, as well as for its stimulation of basic fibroblast growth factor expression. It also contains a destruction box (D box) that is required for its degradation by the APC. The acidic C-terminal region of the encoded protein can act as a transactivation domain. The gene product is mainly a cytosolic protein, although it partially localizes in the nucleus.
Spezifikation
| Securin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Pttg1 | |
| AW555095; C87862; EAP1; Esp1-associated protein; ESP1-associated protein 1; HPTTG; MGC126883; MGC138276; Pituitary tumor transforming; pituitary tumor-transforming 1; pituitary tumor-transforming gene 1; pituitary tumor-transforming gene 1 protein; PTTG; PTTG1; Pttg3; Securin; tumor-transforming protein 1; TUTR1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 30939, 9232 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1495 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95997, Q9CQJ7 | |
| Pttg1 | |
| A synthesized peptide derived from human Securin (aa 100-149). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |