missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA3B Monoclonal Antibody (904201)

Rat Monoclonal Antibody
Marke: Invitrogen™ MA561556
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis.
Spezifikation
| SEMA3B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q62177 | |
| SEMA3B | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 3B , Pro26-Pro722. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 904201 | |
| Unconjugated | |
| SEMA3B | |
| LUCA-1; Seam V antibody; SemA; sema A; sema A(V); sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B; Sema V; SEMA3B; SEMA5; SEMAA; semaphorin 3B; semaphorin A; semaphorin-3B; semaphorin-A; semaphorin-V; semaV | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20347 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur