Learn More
SERPINB4 Antibody (8B8B2), Biotin , Novus Biologicals™
Artikelnummer. 18606385 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18606385 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18606385 Lieferant Novus Biologicals Lieferanten-Nr. NBP305885B
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
SERPINB4 Monoclonal antibody specifically detects SERPINB4 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen SERPINB4
Anwendungen ELISA
Klassifikation Monoclonal
Klon 8B8B2
Konjugat Biotin
Zusammensetzung PBS
Gen-Alias Leupin, Peptidase inhibitor 11, PI-11, PI11clade B (ovalbumin), member 4, serpin peptidase inhibitor, clade B (ovalbumin), member 4, squamous cell carcinoma antigen 1, Squamous cell carcinoma antigen 2
Wirtsspezies Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SERPINB4 (P48594; Met 1-Pro 390).
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Immunology
Primär oder sekundär Primary
Gen-ID (Entrez) 6318
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG